Assay Details
| Gene Symbol: RBAK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005621128.3 | XP_005621185.1 | 2447 | 114 | ||
| XM_022419743.1 | XP_022275451.1 | 2183 | 114 | |||
| XM_022419744.1 | XP_022275452.1 | 2614 | 114 | |||
| XM_022419745.1 | XP_022275453.1 | 2192 | 114 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

RBAK
Genomic Map