Assay Details
Gene Symbol: FBXW4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005637642.3 | XP_005637699.3 | 4 - 5 | 992 | 118 | |
XM_022411670.1 | XP_022267378.1 | 4 - 5 | 428 | 118 | ||
XM_543981.6 | XP_543981.5 | 4 - 5 | 991 | 118 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() |
