Assay Details
| Gene Symbol: PRRG3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005641951.3 | XP_005642008.1 | 4 | 409 | 82 | |
| XM_005641952.1 | XP_005642009.1 | 4 | 342 | 82 | ||
| XM_005641954.3 | XP_005642011.1 | 3 | 216 | 82 | ||
| XM_005641956.1 | XP_005642013.1 | 3 | 131 | 82 | ||
| XM_014111811.2 | XP_013967286.1 | 4 | 1083 | 82 | ||
| XM_014111813.2 | XP_013967288.1 | 3 | 890 | 82 | ||
| XM_022415737.1 | XP_022271445.1 | 3 | 939 | 82 | ||
| XM_022415738.1 | XP_022271446.1 | 4 | 1336 | 82 | ||
| XM_549337.6 | XP_549337.3 | 3 | 191 | 82 | ||
More Information
Set Membership: |
|

PRRG3
Genomic Map