Assay Details
| Gene Symbol: PRR15L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005624609.3 | XP_005624666.1 | 335 | 166 | ||
| XM_845857.5 | XP_850950.2 | 455 | 166 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

PRR15L
Genomic Map