Assay Details
| Gene Symbol: CPSF7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005631664.3 | XP_005631721.1 | 3 - 4 | 446 | 145 | |
| XM_005631665.2 | XP_005631722.1 | 3 - 4 | 479 | 145 | ||
| XM_005631666.3 | XP_005631723.1 | 3 - 4 | 446 | 145 | ||
| XM_005631667.3 | XP_005631724.1 | 3 - 4 | 446 | 145 | ||
| XM_014121128.2 | XP_013976603.1 | 3 - 4 | 446 | 145 | ||
| XM_014121129.2 | XP_013976604.1 | 3 - 4 | 434 | 145 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

CPSF7
Genomic Map