Assay Details
| Gene Symbol: SLC25A21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022421940.1 | XP_022277648.1 | 5 - 6 | 757 | 139 | |
| XM_022421941.1 | XP_022277649.1 | 4 - 5 | 673 | 139 | ||
| XM_022421942.1 | XP_022277650.1 | 6 - 7 | 878 | 139 | ||
| XM_022421943.1 | XP_022277651.1 | 5 - 6 | 300 | 139 | ||
| XM_022421944.1 | XP_022277652.1 | 5 - 6 | 374 | 139 | ||
| XM_022421945.1 | XP_022277653.1 | 5 - 6 | 764 | 139 | ||
| XM_547777.6 | XP_547777.4 | 5 - 6 | 755 | 139 | ||
More Information
Set Membership: |
|

SLC25A21
Genomic Map