Assay Details
| Gene Symbol: CCDC120 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014111890.2 | XP_013967365.1 | 4 - 5 | 1322 | 80 | |
| XM_022415916.1 | XP_022271624.1 | 5 - 6 | 1392 | 80 | ||
| XM_022415917.1 | XP_022271625.1 | 5 - 6 | 1143 | 80 | ||
| XM_022415918.1 | XP_022271626.1 | 4 - 5 | 1073 | 80 | ||
| XM_022415919.1 | XP_022271627.1 | 3 - 4 | 361 | 80 | ||
| XM_022415920.1 | XP_022271628.1 | 5 - 6 | 1335 | 80 | ||
| XM_022415921.1 | XP_022271629.1 | 4 - 5 | 1265 | 80 | ||
| XM_022415922.1 | XP_022271630.1 | 5 - 6 | 1086 | 80 | ||
| XM_022415923.1 | XP_022271631.1 | 4 - 5 | 1016 | 80 | ||
| XM_022415924.1 | XP_022271632.1 | 4 - 5 | 1301 | 80 | ||
More Information
Set Membership: |
|

CCDC120
Genomic Map