Assay Details
| Gene Symbol: RIPOR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022409524.1 | XP_022265232.1 | 10 - 11 | 1733 | 146 | |
| XM_022409525.1 | XP_022265233.1 | 7 - 8 | 662 | 146 | ||
| XM_022409526.1 | XP_022265234.1 | 10 - 11 | 1725 | 146 | ||
| XM_022409527.1 | XP_022265235.1 | 10 - 11 | 1669 | 146 | ||
| XM_022409528.1 | XP_022265236.1 | 9 - 10 | 1549 | 146 | ||
| XM_022409529.1 | XP_022265237.1 | 8 - 9 | 1413 | 146 | ||
| XM_022409530.1 | XP_022265238.1 | 7 - 8 | 1279 | 146 | ||
| XM_022409531.1 | XP_022265239.1 | 8 - 9 | 1405 | 146 | ||
| XM_022409532.1 | XP_022265240.1 | 10 - 11 | 1729 | 146 | ||
| XM_022409533.1 | XP_022265241.1 | 6 - 7 | 921 | 146 | ||
More Information
Set Membership: |
|

RIPOR3
Genomic Map