Assay Details
| Gene Symbol: SNTB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022419277.1 | XP_022274985.1 | 2 | 777 | 73 | |
| XM_536806.6 | XP_536806.5 | 2 | 771 | 73 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

SNTB2
Genomic Map