Assay Details
| Gene Symbol: JOSD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005616264.2 | XP_005616321.1 | 3 | 531 | 123 | |
| XM_005616265.2 | XP_005616322.1 | 3 | 572 | 123 | ||
| XM_005616266.3 | XP_005616323.1 | 3 | 576 | 123 | ||
| XM_849567.4 | XP_854660.2 | 3 | 525 | 123 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

JOSD2
Genomic Map