Assay Details
| Gene Symbol: LGI4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022403832.1 | XP_022259540.1 | 8 - 9 | 2769 | 96 | |
| XM_541696.6 | XP_541696.2 | 8 - 9 | 2651 | 96 | ||
More Information
Set Membership: |
|

LGI4
Genomic Map