Assay Details
| Gene Symbol: ELOVL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005637654.3 | XP_005637711.1 | 4 | 1115 | 85 | |
| XM_005637655.3 | XP_005637712.1 | 4 | 841 | 85 | ||
| XM_005637656.3 | XP_005637713.1 | 4 | 1179 | 85 | ||
More Information
Set Membership: |
|

ELOVL3
Genomic Map