Assay Details
| Gene Symbol: ZDHHC8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110770.1 | NP_001104240.1 | 10 | 1860 | 137 | |
| XM_005636540.3 | XP_005636597.1 | 10 | 1860 | 137 | ||
| XM_005636541.2 | XP_005636598.1 | 10 | 1860 | 137 | ||
| GenBank mRNA | AY871202.1 | - | 10 | 1860 | 137 | |
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

ZDHHC8
Genomic Map