Assay Details
| Gene Symbol: KHDRBS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014118097.2 | XP_013973572.1 | 2 - 3 | 446 | 154 | |
| XM_022426185.1 | XP_022281893.1 | 2 - 3 | 448 | 154 | ||
More Information
Set Membership: |
|

KHDRBS2
Genomic Map