Assay Details
| Gene Symbol: CHST13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005632063.1 | XP_005632120.1 | 536 | 110 | ||
| XM_005632064.1 | XP_005632121.1 | 365 | 110 | |||
More Information
Set Membership: |
|

CHST13
Genomic Map