Assay Details
| Gene Symbol: IL4I1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005616282.2 | XP_005616339.1 | 4 - 5 | 650 | 164 | |
| XM_014118753.2 | XP_013974228.1 | 3 - 4 | 308 | 164 | ||
| XM_022423581.1 | XP_022279289.1 | 4 - 5 | 712 | 164 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

IL4I1
Genomic Map