Assay Details
| Gene Symbol: SLC16A11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005619947.2 | XP_005620004.1 | 3 | 433 | 95 | |
| XM_005619949.3 | XP_005620006.1 | 2 | 192 | 95 | ||
| XM_005619950.2 | XP_005620007.1 | 3 | 559 | 95 | ||
| XM_005619951.2 | XP_005620008.1 | 3 | 505 | 95 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005619949.3 transcripts.
Set Membership: |
|

SLC16A11
Genomic Map