Assay Details
| Gene Symbol: SETD9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022409596.1 | XP_022265304.1 | 2 | 488 | 148 | |
| XM_844651.5 | XP_849744.1 | 2 | 488 | 148 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: SETD9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022409596.1 | XP_022265304.1 | 2 | 488 | 148 | |
| XM_844651.5 | XP_849744.1 | 2 | 488 | 148 | ||
Set Membership: |
|