Assay Details
| Gene Symbol: HOXA10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206432.1 | NP_001193361.1 | 2 | 1127 | 84 | |
| GenBank mRNA | EU332994.1 | - | 2 | 196 | 84 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: HOXA10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206432.1 | NP_001193361.1 | 2 | 1127 | 84 | |
| GenBank mRNA | EU332994.1 | - | 2 | 196 | 84 | |
Set Membership: |
|