Assay Details
| Gene Symbol: ANAPC16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022417545.1 | XP_022273253.1 | 4 | 399 | 124 | |
| XM_022417546.1 | XP_022273254.1 | 4 | 389 | 124 | ||
| XM_022417547.1 | XP_022273255.1 | 4 | 404 | 124 | ||
| XM_856703.5 | XP_861796.1 | 4 | 391 | 124 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

ANAPC16
Genomic Map