Assay Details
| Gene Symbol: YJEFN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022406818.1 | XP_022262526.1 | 2 | 647 | 80 | |
| XM_541921.5 | XP_541921.3 | 3 | 303 | 80 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

YJEFN3
Genomic Map