Assay Details
| Gene Symbol: LOC608110 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022406615.1 | XP_022262323.1 | 4 - 5 | 3096 | 123 | |
| XM_022406616.1 | XP_022262324.1 | 5 - 6 | 487 | 123 | ||
More Information
Set Membership: |
|

LOC608110
Genomic Map