Assay Details
Gene Symbol: USF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005640877.3 | XP_005640934.1 | 7 | 912 | 160 | |
XM_005640878.2 | XP_005640935.1 | 5 | 520 | 160 | ||
XM_545763.6 | XP_545763.2 | 7 | 796 | 160 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
