Assay Details
| Gene Symbol: TMEM41B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005633659.3 | XP_005633716.1 | 3 - 4 | 350 | 112 | |
| XM_022407831.1 | XP_022263539.1 | 4 - 5 | 458 | 112 | ||
| XM_846328.5 | XP_851421.1 | 3 - 4 | 405 | 112 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

TMEM41B
Genomic Map