Assay Details
| Gene Symbol: LOC481202 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014117278.2 | XP_013972753.1 | 10 - 11 | 1002 | 137 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC481202| Gene Symbol: LOC481202 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014117278.2 | XP_013972753.1 | 10 - 11 | 1002 | 137 | |
Set Membership: |
|