Assay Details
| Gene Symbol: TUBD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003024.1 | NP_001003024.1 | 3 - 4 | 408 | 140 | |
| XM_005624321.3 | XP_005624378.2 | 3 - 4 | 537 | 140 | ||
| XM_005624322.3 | XP_005624379.1 | 3 - 4 | 514 | 140 | ||
| XM_014116460.2 | XP_013971935.2 | 3 - 4 | 519 | 140 | ||
| XM_022422519.1 | XP_022278227.1 | 3 - 4 | 514 | 140 | ||
| XM_022422520.1 | XP_022278228.1 | 3 - 4 | 449 | 140 | ||
| XM_022422521.1 | XP_022278229.1 | 3 - 4 | 514 | 140 | ||
| XM_022422522.1 | XP_022278230.1 | 3 - 4 | 514 | 140 | ||
| GenBank mRNA | AF416724.1 | - | 3 - 4 | 408 | 140 | |
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

TUBD1
Genomic Map