Assay Details
| Gene Symbol: ZNF444 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005616098.2 | XP_005616155.1 | 4 | 939 | 62 | |
| XM_005616099.3 | XP_005616156.1 | 4 | 1790 | 62 | ||
| XM_005616100.3 | XP_005616157.1 | 4 | 939 | 62 | ||
| XM_014114758.2 | XP_013970233.1 | 4 | 1141 | 62 | ||
| XM_014114761.2 | XP_013970236.1 | 4 | 908 | 62 | ||
| XM_022422706.1 | XP_022278414.1 | 4 | 1158 | 62 | ||
| XM_022422712.1 | XP_022278420.1 | 4 | 1216 | 62 | ||
More Information
Set Membership: |
|

ZNF444
Genomic Map