Assay Details
| Gene Symbol: TBC1D2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005626858.3 | XP_005626915.1 | 1 - 2 | 525 | 146 | |
| XM_022425644.1 | XP_022281352.1 | 1 - 2 | 525 | 146 | ||
More Information
Set Membership: |
|

TBC1D2
Genomic Map