Assay Details
| Gene Symbol: KDM6A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001197185.1 | NP_001184114.1 | 7 - 8 | 987 | 89 | |
| NM_001197186.1 | NP_001184115.1 | 7 - 8 | 987 | 89 | ||
| XM_005641004.3 | XP_005641061.1 | 7 - 8 | 816 | 89 | ||
| XM_005641005.3 | XP_005641062.1 | 7 - 8 | 816 | 89 | ||
| XM_005641006.3 | XP_005641063.1 | 7 - 8 | 816 | 89 | ||
| GenBank mRNA | AY530157.1 | - | 7 - 8 | 617 | 89 | |
More Information
Set Membership: |
|

KDM6A
Genomic Map