Assay Details
| Gene Symbol: CCR9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284476.1 | NP_001271405.1 | 3 | 1077 | 119 | |
| GenBank mRNA | AB622806.1 | - | 3 | 1077 | 119 | |
More Information
Set Membership: |
|

CCR9
Genomic Map