Assay Details
| Gene Symbol: ARID5B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005618912.3 | XP_005618969.1 | 4 - 5 | 957 | 101 | |
| XM_022417457.1 | XP_022273165.1 | 4 - 5 | 876 | 101 | ||
| XM_022417458.1 | XP_022273166.1 | 4 - 5 | 615 | 101 | ||
| XM_844500.5 | XP_849593.1 | 7 - 8 | 1557 | 101 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

ARID5B
Genomic Map