Assay Details
| Gene Symbol: FGF5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014109888.2 | XP_013965363.1 | 1 - 2 | 74 | ||
| GenBank mRNA | DQ268833.1 | - | 1 - 2 | 375 | 74 | |
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

FGF5
Genomic Map