Assay Details
| Gene Symbol: KLHL32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022426370.1 | XP_022282078.1 | 4 - 5 | 459 | 154 | |
| XM_022426372.1 | XP_022282080.1 | 4 - 5 | 459 | 154 | ||
| XM_022426373.1 | XP_022282081.1 | 2 - 3 | 267 | 154 | ||
| XM_022426374.1 | XP_022282082.1 | 4 - 5 | 459 | 154 | ||
| XM_022426375.1 | XP_022282083.1 | 4 - 5 | 459 | 154 | ||
| XM_022426376.1 | XP_022282084.1 | 4 - 5 | 459 | 154 | ||
More Information
Set Membership: |
|

KLHL32
Genomic Map