Assay Details
| Gene Symbol: ASB15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014119079.2 | XP_013974554.1 | 6 - 7 | 976 | 168 | |
| XM_022427283.1 | XP_022282991.1 | 6 - 7 | 976 | 168 | ||
More Information
Set Membership: |
|

ASB15
Genomic Map