Assay Details
| Gene Symbol: MAPK14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003206.1 | NP_001003206.1 | 7 - 8 | 608 | 84 | |
| XM_005627027.3 | XP_005627084.1 | 7 - 8 | 992 | 84 | ||
| XM_014118022.2 | XP_013973497.1 | 7 - 8 | 998 | 84 | ||
| GenBank mRNA | AF003597.1 | - | 7 - 8 | 608 | 84 | |
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

MAPK14
Genomic Map