Assay Details
| Gene Symbol: HRAS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001287069.1 | NP_001273998.1 | 2 - 3 | 343 | 105 | |
| NM_001287070.1 | NP_001273999.1 | 2 - 3 | 343 | 105 | ||
More Information
Set Membership: |
|

HRAS
Genomic Map