Assay Details
| Gene Symbol: METTL2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005624229.3 | XP_005624286.1 | 161 | 109 | ||
| XM_005624230.2 | XP_005624287.1 | 142 | 109 | |||
| XM_014116390.2 | XP_013971865.1 | 161 | 109 | |||
| XM_022422804.1 | XP_022278512.1 | 161 | 109 | |||
| XM_022422805.1 | XP_022278513.1 | 161 | 109 | |||
| XM_022422806.1 | XP_022278514.1 | 161 | 109 | |||
| XM_537604.6 | XP_537604.3 | 161 | 109 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005624230.2 transcripts.
Set Membership: |
|

METTL2A
Genomic Map