Assay Details
| Gene Symbol: NKPD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005616450.2 | XP_005616507.1 | 5 | 1764 | 70 | |
| XM_022424925.1 | XP_022280633.1 | 4 | 2414 | 70 | ||
More Information
Set Membership: |
|

NKPD1
Genomic Map