Assay Details
| Gene Symbol: MDFIC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005628748.3 | XP_005628805.1 | 4 | 544 | 146 | |
| XM_014119140.2 | XP_013974615.1 | 4 | 577 | 146 | ||
| XM_022427620.1 | XP_022283328.1 | 3 | 325 | 146 | ||
More Information
Set Membership: |
|

MDFIC
Genomic Map