Assay Details
| Gene Symbol: PATZ1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003433403.4 | XP_003433451.1 | 1 | 1496 | 178 | |
| XM_003433404.4 | XP_003433452.1 | 1 | 1496 | 178 | ||
| XM_014107794.2 | XP_013963269.1 | 1 | 1496 | 178 | ||
| XM_022410626.1 | XP_022266334.1 | 1 | 1496 | 178 | ||
| XM_861131.5 | XP_866224.1 | 1 | 1496 | 178 | ||
| XM_861146.5 | XP_866239.1 | 1 | 1496 | 178 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

PATZ1
Genomic Map