Assay Details
| Gene Symbol: CDCP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005632641.3 | XP_005632698.1 | 4 | 849 | 84 | |
| XM_005632642.3 | XP_005632699.1 | 4 | 851 | 84 | ||
More Information
Set Membership: |
|

CDCP1
Genomic Map