Assay Details
| Gene Symbol: AP2M1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001287104.1 | NP_001274033.1 | 10 - 11 | 1228 | 85 | |
| XM_853500.5 | XP_858593.1 | 9 - 10 | 1258 | 85 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

AP2M1
Genomic Map