Assay Details
| Gene Symbol: GNAI2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003364.1 | NP_001003364.1 | 6 - 7 | 761 | 115 | |
| GenBank mRNA | AY563036.1 | - | 6 - 7 | 761 | 115 | |
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

GNAI2
Genomic Map