Assay Details
| Gene Symbol: SERPINE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001197095.1 | NP_001184024.1 | 1173 | 78 | ||
| XM_005620896.3 | XP_005620953.1 | 1340 | 78 | |||
| XM_014114124.2 | XP_013969599.1 | 1378 | 78 | |||
| GenBank mRNA | DN399379.1 | - | 316 | 78 | ||
| DN402495.1 | - | 148 | 78 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

SERPINE1
Genomic Map