Assay Details
| Gene Symbol: CCL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003297.1 | NP_001003297.1 | 2 - 3 | 191 | 70 | |
| GenBank mRNA | U29653.1 | - | 2 - 3 | 191 | 70 | |
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

CCL2
Genomic Map