Assay Details
| Gene Symbol: CHRM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005629578.3 | XP_005629635.1 | 1421 | 107 | ||
| GenBank mRNA | AF084483.1 | - | 322 | 107 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: CHRM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005629578.3 | XP_005629635.1 | 1421 | 107 | ||
| GenBank mRNA | AF084483.1 | - | 322 | 107 | ||
Set Membership: |
|