Assay Details
| Gene Symbol: UGT1A6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003078.1 | NP_001003078.1 | 4 - 5 | 1294 | 68 | |
| XM_005635599.3 | XP_005635656.1 | 4 - 5 | 1332 | 68 | ||
| XM_005635600.3 | XP_005635657.1 | 4 - 5 | 1364 | 68 | ||
| XM_014107454.2 | XP_013962929.1 | 4 - 5 | 1329 | 68 | ||
| GenBank mRNA | AJ290948.1 | - | 4 - 5 | 1294 | 68 | |
More Information
Set Membership: |
|

UGT1A6
Genomic Map