Assay Details
| Gene Symbol: DLA-64 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014378.1 | NP_001014400.1 | 2 - 3 | 350 | 68 | |
| GenBank mRNA | LC171439.1 | - | 2 - 3 | 350 | 68 | |
| LC171441.1 | - | 2 - 3 | 350 | 68 | ||
| LC171442.1 | - | 2 - 3 | 350 | 68 | ||
| LC171444.1 | - | 2 - 3 | 350 | 68 | ||
| LC171445.1 | - | 2 - 3 | 350 | 68 | ||
More Information
Set Membership: |
|

DLA-64
Genomic Map