Assay Details
| Gene Symbol: ST8SIA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001097548.1 | NP_001091017.1 | 3 - 4 | 422 | 70 | |
| XM_005618263.3 | XP_005618320.1 | 4 - 5 | 616 | 70 | ||
| GenBank mRNA | AM422132.1 | - | 3 - 4 | 422 | 70 | |
More Information
Set Membership: |
|

ST8SIA2
Genomic Map