Assay Details
| Gene Symbol: ODAM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001097551.1 | NP_001091020.1 | 3 - 4 | 166 | 63 | |
| XM_022426505.1 | XP_022282213.1 | 3 - 4 | 171 | 63 | ||
| GenBank mRNA | EF121758.1 | - | 3 - 4 | 166 | 63 | |
More Information
Set Membership: |
|

ODAM
Genomic Map